Assay Details
| Gene Symbol: Pelo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134058.3 | NP_598819.2 | 1 - 2 | 971 | 67 | |
| GenBank mRNA | AF148638.1 | - | 1032 | 67 | ||
| AK146382.1 | - | 1 - 2 | 1001 | 67 | ||
| AK168228.1 | - | 1 - 2 | 1001 | 67 | ||
| AK170635.1 | - | 1 - 2 | 971 | 67 | ||
| BC050209.1 | - | 1 - 2 | 884 | 6431710 | 67 | |
| BC057160.1 | - | 1 - 2 | 772 | 5349483 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
cell proliferation
chromosome organization
cell division
nuclear-transcribed mRNA catabolic process, non-stop decay
nuclear-transcribed mRNA catabolic process, no-go decay
RNA surveillance
nuclease activity
endonuclease activity
protein binding
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
metal ion binding
cell cycle
cell proliferation
chromosome organization
cell division
nuclear-transcribed mRNA catabolic process, non-stop decay
nuclear-transcribed mRNA catabolic process, no-go decay
RNA surveillance
nuclease activity
endonuclease activity
protein binding
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29350214 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29350210 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29350213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29350211 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | AK168228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | AK170635.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | BB767039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | BC050209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | NM_134058.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | AK168228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | AK170635.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | BB767039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | BC050209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P175555 | NM_134058.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163179 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | AK168228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | AK170635.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | BB767039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | BC050209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | NM_134058.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | AK168228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | AK170635.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | BB767039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | BC050209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P175555 | NM_134058.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29350197 | - | - |
| Whole Mouse Genome G4122A | A_51_P175552 | BC057160.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175552 | AF148638.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | AK168228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | AK170635.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | BB767039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | BC050209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | NM_134058.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | AK168228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | AK170635.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | BB767039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | BC050209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175555 | NM_134058.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137840 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10694054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10694055 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10580582 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10963648 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10694064 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10963638 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10489492 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10694063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10580581 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10580580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29350211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29350214 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29350213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29350210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29350284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10563453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10643692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10643681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10533865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10771057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10580581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10694054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10643699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10771055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11033933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10986524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10447141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10533866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10580580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10489494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29350265 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10412330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10412331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10412332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10412329 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434071_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10412328 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10407316 | - | - |
| Mouse Genome 430 2.0 Array | 1434071_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434071_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105083_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105083_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300018407.mm.1 | NM_134058.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300018407.mm.1 | NM_134058.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300018407.mm.1 | TC1300002717.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434071_a_at | - | - |
| Mouse Expression Set 430 | 1434071_PM_a_at | - | - |
| Murine Genome U74v2 Set | 161280_r_at | - | - |
| Murine Genome U74v2 Set | 98017_at | - | - |
| Murine Genome U74v2 Set | 161280_PM_r_at | - | - |
| Murine Genome U74v2 Set | 98017_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434071_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434071_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434071_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434071_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3440706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3440706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2676984 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2676984 | - | - |
| mouse-6 v1.1 (Illumina) | scl0105083.1_137-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0105083.1_137-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2853658 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2853658 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0105083.1_137-S | - | - |

Pelo
Genomic Map