Assay Details
| Gene Symbol: Rbm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144948.5 | NP_659197.2 | 4 - 5 | 515 | 101 | |
| NR_037589.1 | 5 - 6 | 620 | 101 | |||
| GenBank mRNA | AF458961.1 | - | 4 - 5 | 101 | ||
| AK005200.1 | - | 4 - 5 | 491 | 101 | ||
| AK006755.1 | - | 4 - 5 | 480 | 101 | ||
| AK008645.1 | - | 4 - 5 | 475 | 101 | ||
| AK011223.1 | - | 4 - 5 | 497 | 101 | ||
| AK015662.1 | - | 4 - 5 | 516 | 101 | ||
| AK151056.1 | - | 4 - 5 | 494 | 101 | ||
| AK153504.1 | - | 5 - 6 | 584 | 101 | ||
| BC054774.1 | - | 4 - 5 | 482 | 6807637 | 101 | |
| BC116931.1 | - | 4 - 5 | 464 | 40126250 | 101 | |
| BC119038.1 | - | 4 - 5 | 464 | 8733787 | 101 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbm7
Genomic Map