Assay Details
| Gene Symbol: Ceacam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039187.1 | NP_001034276.1 | 2 - 3 | 518 | 100 | |
| NM_011926.2 | NP_036056.2 | 2 - 3 | 518 | 100 | ||
| GenBank mRNA | AB236328.2 | - | 2 - 3 | 427 | 100 | |
| AB236329.1 | - | 2 - 3 | 427 | 100 | ||
| AB236331.2 | - | 2 - 3 | 427 | 100 | ||
| AK085408.1 | - | 641 | 100 | |||
| X67278.1 | - | 2 - 3 | 530 | 100 | ||
| X67282.1 | - | 2 - 3 | 530 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ceacam1
Genomic Map