Assay Details
| Gene Symbol: Ceacam9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011927.4 | NP_036057.1 | 1 - 2 | 98 | 112 | |
| GenBank mRNA | AK049543.1 | - | 1 - 2 | 70 | 112 | |
| L49180.1 | - | 1 - 2 | 98 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ceacam9
Genomic Map