Assay Details
| Gene Symbol: Psmb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011971.4 | NP_036101.1 | 2 - 3 | 244 | 78 | |
| GenBank mRNA | AF060092.1 | - | 2 - 3 | 206 | 78 | |
| AK002331.1 | - | 2 - 3 | 245 | 78 | ||
| AK010488.1 | - | 2 - 3 | 225 | 78 | ||
| AK050108.1 | - | 2 - 3 | 242 | 78 | ||
| AK080755.1 | - | 2 - 3 | 244 | 78 | ||
| AK153162.1 | - | 2 - 3 | 277 | 78 | ||
| BC014783.1 | - | 2 - 3 | 211 | 3500033 | 78 | |
| BC098233.1 | - | 1 - 2 | 206 | 1547611 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psmb3
Genomic Map