Assay Details
| Gene Symbol: Psmb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011971.4 | NP_036101.1 | 5 - 6 | 626 | 94 | |
| GenBank mRNA | AK002331.1 | - | 5 - 6 | 627 | 94 | |
| AK010488.1 | - | 5 - 6 | 607 | 94 | ||
| AK153162.1 | - | 5 - 6 | 659 | 94 | ||
| BC014783.1 | - | 5 - 6 | 593 | 3500033 | 94 | |
| BC098233.1 | - | 4 - 5 | 588 | 1547611 | 94 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
proteasomal ubiquitin-independent protein catabolic process
protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
proteolysis
proteasomal ubiquitin-independent protein catabolic process
protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | AK010488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | AK153162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | BC098233.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | NM_011971.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | AK002331.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | AK010488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | AK153162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | BC098233.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | NM_011971.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P9437 | AK002331.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780908 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53211 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45024 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159228 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137529 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168723 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | AK010488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | AK153162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | BC098233.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | NM_011971.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | AK002331.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | AK010488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | AK153162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | BC098233.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | NM_011971.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P9437 | AK002331.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209193 | AK010488.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209193 | AK153162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209193 | BC014783.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209193 | BC098233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209193 | NM_011971.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | AK010488.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | AK153162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | BC098233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | NM_011971.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | AK002331.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | AK010488.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | AK153162.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | BC098233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | NM_011971.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P9437 | AK002331.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28052212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28052219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28052217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28052215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28052211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28052224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28052216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28052222 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137529 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168723 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07456394 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159228 | - | - |
| Whole Mouse Genome G4122A | A_52_P9437 | AK010488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | AK153162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | BC098233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | NM_011971.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | AK002331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | AK010488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | AK153162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | BC098233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | NM_011971.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9437 | AK002331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P209193 | AK010488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P209193 | AK153162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P209193 | BC014783.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P209193 | BC098233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P209193 | NM_011971.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121029 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131934 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159228 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168723 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08986595 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08986592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28052152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09591430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09053665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09283392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08986610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09020125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09332437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09128316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28052206 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434644 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417052_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460198_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380815 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10434643 | - | - |
| Mouse Genome 430 2.0 Array | 1417052_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460198_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417052_at | - | - |
| Mouse Genome 430 2.0 Array | 1460198_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100014269.mm.1 | NM_011971.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014269.mm.1 | NM_011971.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014269.mm.1 | TC1100001546.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417052_at | - | - |
| Mouse Expression Set 430 | 1460198_a_at | - | - |
| Mouse Expression Set 430 | 1417052_PM_at | - | - |
| Mouse Expression Set 430 | 1460198_PM_a_at | - | - |
| Murine Genome U74v2 Set | 94025_at | - | - |
| Murine Genome U74v2 Set | 94025_PM_at | - | - |
| Murine Genome U74v2 Set | 171521_PM_at | - | - |
| Murine Genome U74v2 Set | 171521_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460198_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460198_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417052_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417052_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417052_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460198_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460198_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417052_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 896064 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106110162 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4280594 | - | - |
| MouseWG-6 v2 Expression BeadChip | 107100397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106110162 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107100397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4280594 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708599 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235121 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2430164 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2708599 | - | - |
| mouse-6 v1.1 (Illumina) | scl40954.3.1_30-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_31981321-S | - | - |
| mouse-6 v1.1 (Illumina) | scl026446.4_108-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280594 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106110162 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107100397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl026446.4_108-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280594 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106110162 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107100397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2708599 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2430164 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708599 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl026446.4_108-S | - | - |

Psmb3
Genomic Map