Assay Details
| Gene Symbol: Klra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016659.3 | NP_057868.2 | 1 - 2 | 44 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_016659.3 transcripts.
Set Membership: |
|

Klra1
Genomic Map