Assay Details
Gene Symbol: Olfr1269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146342.1 | NP_666454.1 | 1 | 748 | 84 | |
GenBank mRNA | BC116956.1 | - | 1 | 769 | 40126275 | 84 |
BC116958.1 | - | 1 | 769 | 40126277 | 84 |
Gene Symbol: Olfr1265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146343.1 | NP_666455.1 | 1 | 748 | 84 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980094 | NM_146343.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988989 | NM_146343.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980094 | NM_146343.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988989 | NM_146343.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P135864 | BC116958.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P135864 | NM_146343.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P135864 | NM_146342.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P135864 | BC116956.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415630 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415636 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415633 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415627 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415647 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415650 | - | - |
Whole Mouse Genome G4122A | A_51_P135864 | BC116958.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P135864 | NM_146343.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P135864 | NM_146342.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P135864 | BC116956.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123385 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106673 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104090 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02204292 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02204290 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02204294 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02204280 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02204274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415932 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415646 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415668 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01228444 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415649 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415937 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02204292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01562403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415970 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415939 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415915 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01562395 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01800040 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02080214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415664 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415650 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415917 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415665 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01828573 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02080222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415950 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02110315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415666 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21415961 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484864 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484864 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484868 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473642 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484863 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484867 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10473641 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484863 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034124.mm.1 | TC0200004080.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 395910 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2320079 | - | - |
MouseWG-6 v2 Expression BeadChip | 870050 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2320079 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 870050 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000870050 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002320079 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2738845 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257124 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257124 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2738845 | - | - |
mouse-6 v1.1 (Illumina) | scl0258340.1_43-S | - | - |
mouse-6 v1.1 (Illumina) | scl0258339.1_42-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2320079 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 870050 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258340.1_43-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258339.1_42-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320079 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 870050 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2738845 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1257124 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2738845 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257124 | - | - |
