Assay Details
| Gene Symbol: Olfr248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146714.2 | NP_666925.2 | 1 | 47 | 94 | |
| GenBank mRNA | BC131960.1 | - | 1 | 47 | 40130237 | 94 |
| BC131962.1 | - | 1 | 47 | 40130239 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC131960.1, BC131962.1, NM_146714.2 transcripts.
Set Membership: |
|

Olfr248
Genomic Map