Assay Details
Gene Symbol: Olfr689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146750.1 | NP_666961.1 | 1 | 654 | 87 | |
GenBank mRNA | BC119379.1 | - | 1 | 678 | 8734128 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970893 | NM_146750.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970893 | BC119379.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170471 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970893 | NM_146750.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970893 | BC119379.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P295007 | NM_146750.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P295007 | BC119379.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170471 | - | - |
Whole Mouse Genome G4122A | A_51_P295007 | NM_146750.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P295007 | BC119379.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136832 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117423 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06668242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391648 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391673 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391634 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06544118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06552723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06427984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25391678 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10555914 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10555916 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10555918 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10555917 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10555913 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10555915 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700012943.mm.1 | TC0700001547.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 898836 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 7100609 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007100609 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2628377 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2628377 | - | - |
mouse-6 v1.1 (Illumina) | scl9376.1.1_38-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7100609 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl9376.1.1_38-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100609 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2628377 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2628377 | - | - |
