Assay Details
Gene Symbol: Olfr912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146810.2 | NP_667021.2 | 1 - 2 | 44 | 138 | |
GenBank mRNA | CB174492.1 | - | 1 - 2 | 44 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_146810.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
