Assay Details
Gene Symbol: Olfr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146923.2 | NP_667134.2 | 1 - 2 | 103 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_146923.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
