Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200034067.mm.1 |
NM_146969.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034067.mm.1 |
NM_146969.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034067.mm.1 |
TC0200004062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034068.mm.1 |
NM_146969.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034068.mm.1 |
NM_146969.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034068.mm.1 |
TC0200004062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034066.mm.1 |
NM_146969.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034066.mm.1 |
NM_146969.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034066.mm.1 |
TC0200004062.mm.1 |
MTA TC |