Assay Details
| Gene Symbol: Olfr623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147122.2 | NP_667333.2 | 1 | 47 | 89 | |
| GenBank mRNA | BC119275.1 | - | 1 | 47 | 8734024 | 89 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC119275.1, NM_147122.2 transcripts.
Set Membership: |
|

Olfr623
Genomic Map