Assay Details
| Gene Symbol: Tagap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147155.2 | NP_671511.2 | 2 - 3 | 184 | 102 | |
| GenBank mRNA | AF314818.1 | - | 2 - 3 | 184 | 102 | |
| AV444953.1 | - | 2 - 3 | 228 | 102 | ||
| BC119550.1 | - | 2 - 3 | 184 | 102 | ||
| BC119551.1 | - | 2 - 3 | 184 | 102 | ||
| BC139797.1 | - | 2 - 3 | 182 | 102 | ||
| BQ555561.1 | - | 1 - 2 | 59 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tagap1
Genomic Map