Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700005197.mm.1 |
NM_153063.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005197.mm.1 |
NM_153063.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005197.mm.1 |
TC1700000571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005195.mm.1 |
NM_153063.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005195.mm.1 |
NM_153063.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005195.mm.1 |
TC1700000571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005196.mm.1 |
NM_153063.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005196.mm.1 |
NM_153063.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005196.mm.1 |
TC1700000571.mm.1 |
MTA TC |