Assay Details
Gene Symbol: Zfp472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153063.3 | NP_694703.2 | 3 - 4 | 283 | 103 | |
GenBank mRNA | AK142395.1 | - | 3 - 4 | 274 | 103 | |
AY195875.1 | - | 3 - 4 | 218 | 103 | ||
BC023090.1 | - | 3 - 4 | 218 | 103 | ||
BC027407.1 | - | 3 - 4 | 273 | 4945483 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
