Assay Details
| Gene Symbol: Klrb1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153094.2 | NP_694734.1 | 1 - 2 | 136 | 92 | |
| NR_024262.1 | 1 - 2 | 136 | 92 | |||
| NR_024263.1 | 1 - 2 | 136 | 92 | |||
| GenBank mRNA | AK041601.1 | - | 1 - 2 | 137 | 92 | |
| AK154278.1 | - | 1 - 2 | 123 | 92 | ||
| AK170023.1 | - | 1 - 2 | 113 | 92 | ||
| AY029597.1 | - | 1 - 2 | 101 | 92 | ||
| BC107177.1 | - | 1 - 2 | 117 | 92 | ||
| BC107178.1 | - | 1 - 2 | 117 | 92 | ||
| BC127141.1 | - | 1 - 2 | 117 | 92 | ||
| BC127142.1 | - | 1 - 2 | 113 | 40050668 | 92 | |
| DQ237931.1 | - | 1 - 2 | 89 | 92 | ||
| HQ713435.1 | - | 1 - 2 | 89 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107177.1, BC107178.1, BC127141.1 transcripts.
Set Membership: |
|

Klrb1f
Genomic Map