Assay Details
| Gene Symbol: Klrb1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153094.2 | NP_694734.1 | 3 - 4 | 305 | 90 | |
| GenBank mRNA | AK041601.1 | - | 3 - 4 | 306 | 90 | |
| AK154278.1 | - | 3 - 4 | 292 | 90 | ||
| AK170023.1 | - | 3 - 4 | 282 | 90 | ||
| AY029597.1 | - | 3 - 4 | 270 | 90 | ||
| BC127142.1 | - | 3 - 4 | 282 | 40050668 | 90 | |
| DQ237931.1 | - | 3 - 4 | 258 | 90 | ||
| HQ713435.1 | - | 3 - 4 | 258 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klrb1f
Genomic Map