Assay Details
| Gene Symbol: Snx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172277.2 | NP_758481.1 | 7 - 8 | 909 | 107 | |
| GenBank mRNA | AK045061.1 | - | 7 - 8 | 902 | 107 | |
| AK082871.1 | - | 7 - 8 | 902 | 107 | ||
| AK148429.1 | - | 7 - 8 | 902 | 107 | ||
| AK156817.1 | - | 7 - 8 | 855 | 107 | ||
| AK161672.1 | - | 6 - 7 | 843 | 107 | ||
| BC019142.1 | - | 6 - 7 | 905 | 107 | ||
| BC037599.1 | - | 7 - 8 | 935 | 5150754 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx8
Genomic Map