Assay Details
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172624.3 | NP_766212.2 | 12 - 13 | 1653 | 77 | |
| GenBank mRNA | AK050021.1 | - | 12 - 13 | 1595 | 77 | |
| AK050023.1 | - | 12 - 13 | 1655 | 77 | ||
| AK078301.1 | - | 12 - 13 | 1601 | 77 | ||
| AK131178.1 | - | 9 - 10 | 3516 | 77 | ||
| AK201854.1 | - | 1 - 2 | 158 | 77 | ||
| AK204395.1 | - | 1 - 2 | 158 | 77 | ||
| BC057631.1 | - | 12 - 13 | 1652 | 4191886 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpp9
Genomic Map