Assay Details
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172624.3 | NP_766212.2 | 17 - 18 | 2325 | 105 | |
| GenBank mRNA | AK050021.1 | - | 17 - 18 | 2267 | 105 | |
| AK050023.1 | - | 17 - 18 | 2327 | 105 | ||
| AK078301.1 | - | 17 - 18 | 2273 | 105 | ||
| AK131178.1 | - | 14 - 15 | 4188 | 105 | ||
| BC057631.1 | - | 17 - 18 | 2324 | 4191886 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpp9
Genomic Map