Assay Details
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172624.3 | NP_766212.2 | 1 - 2 | 147 | 77 | |
| GenBank mRNA | AK039652.1 | - | 1 - 2 | 165 | 77 | |
| AK050021.1 | - | 1 - 2 | 89 | 77 | ||
| AK050023.1 | - | 1 - 2 | 148 | 77 | ||
| AK078301.1 | - | 1 - 2 | 95 | 77 | ||
| BC057631.1 | - | 1 - 2 | 146 | 4191886 | 77 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK050021.1,AK078301.1,BC057631.1,NM_172624.3 transcripts.
Set Membership: |
|

Dpp9
Genomic Map