Assay Details
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172624.3 | NP_766212.2 | 20 - 21 | 2729 | 80 | |
| GenBank mRNA | AK050021.1 | - | 20 - 21 | 2671 | 80 | |
| AK050023.1 | - | 20 - 21 | 2731 | 80 | ||
| AK078301.1 | - | 20 - 21 | 2677 | 80 | ||
| AK131178.1 | - | 18 - 19 | 4712 | 80 | ||
| BC057631.1 | - | 20 - 21 | 2728 | 4191886 | 80 | |
More Information
Set Membership: |
|

Dpp9
Genomic Map