Assay Details
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172624.3 | NP_766212.2 | 3 - 4 | 458 | 74 | |
| GenBank mRNA | AK039652.1 | - | 3 - 4 | 476 | 74 | |
| AK050021.1 | - | 3 - 4 | 400 | 74 | ||
| AK050023.1 | - | 3 - 4 | 459 | 74 | ||
| AK078301.1 | - | 3 - 4 | 406 | 74 | ||
| BC057631.1 | - | 3 - 4 | 457 | 4191886 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpp9
Genomic Map