Assay Details
| Gene Symbol: Prpf38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172697.3 | NP_766285.2 | 1 - 2 | 349 | 89 | |
| GenBank mRNA | AK075925.1 | - | 1 - 2 | 341 | 89 | |
| AK135431.1 | - | 1 - 2 | 142 | 89 | ||
| AK169741.1 | - | 1 - 2 | 302 | 89 | ||
| BC024734.1 | - | 1 - 2 | 322 | 89 | ||
| BC025853.1 | - | 1 - 2 | 251 | 89 | ||
| BC139277.1 | - | 1 - 2 | 287 | 8862299 | 89 | |
| BC139305.1 | - | 1 - 2 | 285 | 8862327 | 89 | |
| BY283675.1 | - | 1 - 2 | 349 | 89 | ||
| CT010186.1 | - | 1 - 2 | 127 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prpf38a
Genomic Map