Assay Details
| Gene Symbol: Prpf38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172697.3 | NP_766285.2 | 3 - 4 | 631 | 108 | |
| GenBank mRNA | AK075925.1 | - | 3 - 4 | 623 | 108 | |
| AK169741.1 | - | 3 - 4 | 584 | 108 | ||
| BC024734.1 | - | 3 - 4 | 604 | 108 | ||
| BC025853.1 | - | 3 - 4 | 533 | 108 | ||
| BC139277.1 | - | 3 - 4 | 569 | 8862299 | 108 | |
| BC139305.1 | - | 3 - 4 | 567 | 8862327 | 108 | |
| CT010186.1 | - | 3 - 4 | 409 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf38a
Genomic Map