Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172715.3 | NP_766303.1 | 11 - 12 | 1417 | 94 | |
| GenBank mRNA | AK030171.1 | - | 12 - 13 | 1602 | 94 | |
| AK138410.1 | - | 10 - 11 | 1577 | 94 | ||
| AK160261.1 | - | 11 - 12 | 1362 | 94 | ||
| BC096769.1 | - | 11 - 12 | 30101305 | 94 | ||
| BC138228.1 | - | 10 - 11 | 1170 | 8861247 | 94 | |
| BC145669.1 | - | 10 - 11 | 40130972 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat9
Genomic Map