Assay Details
Gene Symbol: Prune | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173347.2 | NP_775482.1 | 1 - 2 | 491 | 84 | |
GenBank mRNA | AK081637.1 | - | 1 - 2 | 182 | 84 | |
AK142160.1 | - | 1 - 2 | 90 | 84 | ||
BC057546.1 | - | 1 - 2 | 491 | 6416282 | 84 | |
BC058635.1 | - | 1 - 2 | 220 | 5698271 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK081637.1, AK142160.1, BC057546.1, BC058635.1, NM_173347.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
