Assay Details
| Gene Symbol: Taf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175770.4 | NP_786964.1 | 1 - 2 | 254 | 98 | |
| GenBank mRNA | AF144562.1 | - | 1 - 2 | 201 | 98 | |
| AK029380.1 | - | 1 - 2 | 216 | 98 | ||
| AK075700.1 | - | 1 - 2 | 202 | 98 | ||
| AK076362.1 | - | 1 - 2 | 217 | 98 | ||
| AK089054.1 | - | 1 - 2 | 222 | 98 | ||
| BC029673.1 | - | 1 - 2 | 185 | 5373242 | 98 | |
| DV656353.1 | - | 1 - 2 | 254 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF144562.1, AK029380.1, AK075700.1, AK076362.1, AK089054.1, BC029673.1, NM_175770.4 transcripts.
Set Membership: |
|

Taf7
Genomic Map