Assay Details
| Gene Symbol: Hist2h2be | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178214.3 | NP_835586.2 | 1 | 354 | 105 | |
| GenBank mRNA | AK078533.1 | - | 1 | 355 | 105 | |
| AK090189.1 | - | 1 | 357 | 105 | ||
| AK143398.1 | - | 1 | 358 | 105 | ||
| AK162494.1 | - | 1 | 358 | 105 | ||
| AK167546.1 | - | 1 | 357 | 105 | ||
| AK168767.1 | - | 1 | 354 | 105 | ||
| AK170749.1 | - | 1 | 357 | 105 | ||
| BC047137.1 | - | 1 | 334 | 6311571 | 105 | |
| BC058562.1 | - | 1 | 356 | 105 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hist2h2be
Genomic Map