Assay Details
Gene Symbol: BC049715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178776.3 | NP_848891.2 | 2 - 3 | 320 | 91 | |
GenBank mRNA | BC049715.1 | - | 2 - 3 | 320 | 6741857 | 91 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC049715.1,NM_178776.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073965 | AK044622.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073965 | BX638225.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073965 | NM_178776.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134591 | BC049715.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134591 | NM_178776.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183848 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947336 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634437 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678441 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635669 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728622 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160112 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134591 | BC049715.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134591 | NM_178776.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073965 | AK044622.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073965 | BX638225.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073965 | NM_178776.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874665 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874663 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874657 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06081805 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160112 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06081781 | - | - |
Whole Mouse Genome G4122A | A_52_P144138 | AK044622.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118907 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874678 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05306023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05222378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874668 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05222374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05306001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05207444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05222372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05198399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05306011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874663 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05306005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05306025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05870028 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05207447 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10542405 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542406 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542403 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542402 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10542401 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10548871 | - | - |
Mouse Genome 430 2.0 Array | 1457323_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457323_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600012958.mm.1 | TC0600001629.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600012958.mm.1 | NM_178776.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012958.mm.1 | NM_178776.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012955.mm.1 | TC0600001629.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600012955.mm.1 | NM_178776.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012955.mm.1 | NM_178776.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012959.mm.1 | TC0600001629.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600012959.mm.1 | NM_178776.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012959.mm.1 | NM_178776.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012952.mm.1 | TC0600001629.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600012952.mm.1 | NM_178776.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600012952.mm.1 | NM_178776.3 | Refseq |
Murine Genome U74v2 Set | 166357_PM_at | - | - |
Murine Genome U74v2 Set | 166357_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 575879 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5130441 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5130441 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005130441 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777957 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2777957 | - | - |
mouse-6 v1.1 (Illumina) | scl29418.5.1_34-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5130441 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl29418.5.1_34-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130441 | - | - |
