Assay Details
| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205236.1 | NP_001192165.1 | 2 - 3 | 552 | 103 | |
| NM_183288.3 | NP_899111.2 | 2 - 3 | 552 | 103 | ||
| GenBank mRNA | AK017636.1 | - | 2 - 3 | 245 | 103 | |
| AK154926.1 | - | 2 - 3 | 553 | 103 | ||
| CJ068176.1 | - | 2 - 3 | 251 | 103 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC048412.1 | - | 327 | Chr. - - [-] on Build | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017636.1, NM_001205236.1, NM_183288.3 transcripts.
Set Membership: |
|

Arhgap27
Genomic Map