Assay Details
Gene Symbol: Mrgprd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203490.3 | NP_987075.1 | 1 - 2 | 446 | 103 | |
GenBank mRNA | AY762266.1 | - | 1 - 2 | 446 | 103 | |
BC137948.1 | - | 1 - 2 | 352 | 8860967 | 103 | |
BC137950.1 | - | 1 - 2 | 352 | 8860969 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY762266.1, BC137948.1, BC137950.1, NM_203490.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
