Assay Details
| Gene Symbol: Mfap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026220.4 | NP_080496.1 | 7 | 3044 | 84 | |
| GenBank mRNA | AK161000.1 | - | 7 | 3045 | 84 | |
| AK168801.1 | - | 7 | 3050 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21581276 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011126 | AK161000.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011126 | NM_026220.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023864 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023864 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011126 | AK161000.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011126 | NM_026220.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21581279 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04670985 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04670950 | - | - |
| Whole Mouse Genome G4122A | A_51_P229198 | AK018514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P229198 | AK088946.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P229198 | AK169029.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P229198 | NM_001081975.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P229198 | AK003440.1 | mouse_transcriptome |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21581276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21581177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21581178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21581162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21581197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21581166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21581170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21581197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21581194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21581199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01248286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01807314 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10486864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486869 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486860 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486861 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486865 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486862 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486863 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449445_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419370_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486867 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486858 | - | - |
| Mouse Genome 430 2.0 Array | 1449444_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1449444_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1449445_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1419370_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1449445_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1419370_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67532_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67532_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200038117.mm.1 | NM_026220.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038117.mm.1 | NM_026220.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038117.mm.1 | TC0200004492.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449444_PM_a_at | - | - |
| Mouse Expression Set 430 | 1449445_x_at | - | - |
| Mouse Expression Set 430 | 1449444_a_at | - | - |
| Mouse Expression Set 430 | 1419370_PM_a_at | - | - |
| Mouse Expression Set 430 | 1419370_a_at | - | - |
| Mouse Expression Set 430 | 1449445_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449445_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419370_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449444_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419370_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449444_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449445_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6660092 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660092 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2702044 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2702044 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067532.1_134-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660092 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067532.1_134-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660092 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2702044 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2702044 | - | - |

Mfap1a
Genomic Map