Assay Details
| Gene Symbol: Aph1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026674.3 | NP_080950.1 | 1 | 53 | 54 | |
| GenBank mRNA | AK155147.1 | - | 1 | 67 | 54 | |
| BC050923.1 | - | 1 | 53 | 6410104 | 54 | |
| BC063254.1 | - | 1 | 53 | 6410104 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155147.1,BC050923.1,BC063254.1,NM_026674.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aph1c
Genomic Map