Assay Details
| Gene Symbol: Arf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007476.3 | NP_031502.1 | 1 | 56 | 105 | |
| GenBank mRNA | CN705066.1 | - | 1 | 87 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_007476.3 transcripts.
Set Membership: |
|

Arf1
Genomic Map