Assay Details
| Gene Symbol: Rdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009040.3 | NP_033066.1 | 4 | 3049 | 82 | |
| GenBank mRNA | AF030513.1 | - | 4 | 3050 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P240796 | NM_009040.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P240796 | AF030513.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P240796 | NM_009040.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P240796 | AF030513.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 577160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583321 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933583 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262756 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86142 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130890 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148804 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P240796 | NM_009040.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P240796 | AF030513.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P240796 | NM_009040.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P240796 | AF030513.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240796 | NM_009040.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240796 | AF030513.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240796 | NM_009040.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240796 | AF030513.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538140 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148804 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130890 | - | - |
| Whole Mouse Genome G4122A | A_52_P240796 | NM_009040.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240796 | AF030513.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240796 | NM_009040.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240796 | AF030513.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129052 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121921 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130890 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148804 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08701783 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08701794 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08701791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08701806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08701788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08701794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08604013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08577615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08380130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538139 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367049 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367047 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367046 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367048 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420541_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367045 | - | - |
| Mouse Genome 430 2.0 Array | 1420541_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420541_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19683_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19683_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000011351.mm.1 | NM_009040.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011351.mm.1 | NM_009040.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011351.mm.1 | TC1000003221.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420541_PM_at | - | - |
| Mouse Expression Set 430 | 1420541_at | - | - |
| Murine Genome U74v2 Set | 102013_at | - | - |
| Murine Genome U74v2 Set | 102013_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420541_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420541_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420541_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420541_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 508861 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2810427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002810427 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219440 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219440 | - | - |
| mouse-6 v1.1 (Illumina) | scl019683.5_201-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2810427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl019683.5_201-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2810427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219440 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219440 | - | - |

Rdh16
Genomic Map