Assay Details
| Gene Symbol: Srsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079694.1 | NP_001073162.1 | 1 | 47 | 105 | |
| NM_001079695.1 | NP_001073163.1 | 1 | 47 | 105 | ||
| NM_009159.2 | NP_033185.2 | 1 | 47 | 105 | ||
| GenBank mRNA | BU935699.1 | - | 1 | 52 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001079694.1, NM_001079695.1, NM_009159.2 transcripts.
Set Membership: |
|

Srsf5
Genomic Map