Assay Details
| Gene Symbol: Vcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009503.4 | NP_033529.3 | 1 | 50 | 91 | |
| GenBank mRNA | BY732145.1 | - | 1 | 50 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_009503.4 transcripts.
This assay is in the 5'UTR region of the NM_009503.4 transcripts.
Set Membership: |
|

Vcp
Genomic Map