Assay Details
| Gene Symbol: Chil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009892.2 | NP_034022.2 | 47 | 83 | ||
| GenBank mRNA | BY117835.1 | - | 50 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_009892.2 transcripts.
Set Membership: |
|

Chil3
Genomic Map