Assay Details
| Gene Symbol: Tgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054089.4 | NP_473430.3 | 1 | 152 | 101 | |
| GenBank mRNA | AK030953.1 | - | 1 | 144 | 101 | |
| AK044895.1 | - | 1 | 153 | 101 | ||
| AK050044.1 | - | 1 | 82 | 101 | ||
| AK170949.1 | - | 1 | 82 | 101 | ||
| BC058183.1 | - | 1 | 156 | 101 | ||
| BC075728.1 | - | 1 | 133 | 6513964 | 101 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC058183.1,BC075728.1,NM_054089.4 transcripts.
This assay is in the 5'UTR region of the BC058183.1,BC075728.1,NM_054089.4 transcripts.
Set Membership: |
|

Tgs1
Genomic Map