Assay Details
| Gene Symbol: Tubg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134024.2 | NP_598785.1 | 1 | 45 | 77 | |
| GenBank mRNA | AK160125.1 | - | 1 | 58 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_134024.2 transcripts.
This assay is in the 5'UTR region of the NM_134024.2 transcripts.
Set Membership: |
|

Tubg1
Genomic Map