Assay Details
| Gene Symbol: Acot5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145444.3 | NP_663419.3 | 1 | 50 | 100 | |
| GenBank mRNA | BC016076.1 | - | 1 | 51 | 4485973 | 100 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC016076.1,NM_145444.3 transcripts.
This assay is in the 5'UTR region of the BC016076.1,NM_145444.3 transcripts.
Set Membership: |
|

Acot5
Genomic Map