Assay Details
| Gene Symbol: Ttc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177625.4 | NP_808293.1 | 4 | 2151 | 110 | |
| GenBank mRNA | AK081214.1 | - | 4 | 2128 | 110 | |
| AK140574.1 | - | 1 | 923 | 110 | ||
| AW046075.1 | - | 1 | 110 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ttc5
Genomic Map