Assay Details
| Gene Symbol: Orai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178751.3 | NP_848866.2 | 3 | 3648 | 100 | |
| GenBank mRNA | AK042940.1 | - | 3 | 3649 | 100 | |
| BC042668.1 | - | 1 | 1630 | 100 | ||
| BC066070.1 | - | 3 | 3595 | 6853893 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119778 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119777 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119779 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066543 | BC066070.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066543 | BC042668.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066543 | AK042940.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066543 | AK087038.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066543 | NM_178751.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168845 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262685 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658282 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37396 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132942 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174393 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129186 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2066543 | BC066070.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2066543 | BC042668.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2066543 | AK042940.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2066543 | AK087038.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2066543 | NM_178751.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075385 | BC066070.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075385 | BC042668.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075385 | AK042940.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075385 | AK079619.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075385 | AK087038.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24119805 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174393 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01229717 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129186 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01229707 | - | - |
| Whole Mouse Genome G4122A | A_51_P267634 | BC066070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267634 | BC042668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267634 | AK042940.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267634 | NM_178751.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105145 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129186 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174393 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372489 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372488 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372480 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04372484 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04168650 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119777 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119779 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24119778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24119792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04151335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04925393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04838402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119804 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534573 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534572 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434762_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434763_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10436426 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534570 | - | - |
| Mouse Genome 430 2.0 Array | 1434763_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434762_at | - | - |
| Mouse Genome 430 2.0 Array | 1434763_at | - | - |
| Mouse Genome 430 2.0 Array | 1434762_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269717_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269717_PM_at | - | - |
| Mouse Expression Set 430 | 1434763_at | - | - |
| Mouse Expression Set 430 | 1434762_at | - | - |
| Mouse Expression Set 430 | 1434762_PM_at | - | - |
| Mouse Expression Set 430 | 1434763_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434763_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434762_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434763_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434762_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 316945 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5910551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005910551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624293 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2624293 | - | - |
| mouse-6 v1.1 (Illumina) | scl0269717.2_160-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5910551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0269717.2_160-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2624293 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2624293 | - | - |

Orai2
Genomic Map