Assay Details
Gene Symbol: Zfp729a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183146.3 | NP_898969.2 | 4 | 4343 | 105 | |
GenBank mRNA | AK138962.1 | - | 4 | 4343 | 105 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016976 | AK138962.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016976 | NM_183146.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | BC082605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | NM_001001187.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | AB010351.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | AB010374.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | AK032995.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | BC082605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | NM_001001187.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | AB010351.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | AB010374.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P120168 | AK032995.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210901 | BC082605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210901 | NM_001001187.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210901 | AK032995.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210901 | BC082605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210901 | NM_001001187.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210901 | AK032995.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69897 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210901 | BC082605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210901 | NM_001001187.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210901 | AK032995.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210901 | BC082605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210901 | NM_001001187.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210901 | AK032995.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | BC082605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | NM_001001187.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | AB010351.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | AB010374.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | AK032995.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | BC082605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | NM_001001187.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | AB010351.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | AB010374.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P120168 | AK032995.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016976 | AK138962.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016976 | NM_183146.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109437 | - | - |
Whole Mouse Genome G4122A | A_51_P468609 | BC082605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P468609 | NM_001001187.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P468609 | AK032995.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P210901 | BC082605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P210901 | NM_001001187.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P210901 | AK032995.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P210901 | BC082605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P210901 | NM_001001187.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P210901 | AK032995.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | BC082605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | NM_001001187.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | AB010351.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | AB010374.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | AK032995.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | BC082605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | NM_001001187.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | AB010351.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | AB010374.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P120168 | AK032995.1 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10690091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109440 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109448 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109376 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109362 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10829319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109361 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10659905 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10487533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109363 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10617301 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10410359 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10410357 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10410356 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10410358 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10410360 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10405917 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10410361 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1440125_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438491_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1442654_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10405916 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10410355 | - | - |
Mouse Genome 430 2.0 Array | 1440125_at | - | - |
Mouse Genome 430 2.0 Array | 1442654_at | - | - |
Mouse Genome 430 2.0 Array | 1438491_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1442654_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440125_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1438491_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 212281_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 212281_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300014719.mm.1 | NM_183146.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014719.mm.1 | NM_183146.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014719.mm.1 | TC1300002255.mm.1 | MTA TC |
Mouse Expression Set 430 | 1440125_at | - | - |
Mouse Expression Set 430 | 1438491_PM_x_at | - | - |
Mouse Expression Set 430 | 1442654_PM_at | - | - |
Mouse Expression Set 430 | 1438491_x_at | - | - |
Mouse Expression Set 430 | 1440125_PM_at | - | - |
Mouse Expression Set 430 | 1442654_at | - | - |
Murine Genome U74v2 Set | 132806_PM_at | - | - |
Murine Genome U74v2 Set | 132806_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440125_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440125_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438491_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442654_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438491_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442654_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 398926 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6590075 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6590075 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1340168 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006590075 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001340168 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249177 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657930 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2657930 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1249177 | - | - |
mouse-6 v1.1 (Illumina) | scl00212281.1_147-S | - | - |
mouse-6 v1.1 (Illumina) | scl00212281.1_327-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6590075 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1340168 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00212281.1_147-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00212281.1_327-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340168 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590075 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2657929 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2657930 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2657930 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2657929 | - | - |
