Assay Details
| Gene Symbol: Ensa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026212.1 | NP_001021383.1 | 1 | 144 | 69 | |
| NM_019561.2 | NP_062507.1 | 1 | 144 | 69 | ||
| GenBank mRNA | AK006149.1 | - | 1 | 114 | 69 | |
| AK011334.1 | - | 1 | 145 | 69 | ||
| AK019170.1 | - | 1 | 145 | 69 | ||
| AK149726.1 | - | 1 | 112 | 69 | ||
| AK161290.1 | - | 1 | 110 | 69 | ||
| BC020533.1 | - | 1 | 114 | 69 | ||
| BC058802.1 | - | 1 | 93 | 6475839 | 69 | |
| BC059724.1 | - | 1 | 109 | 6773379 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006149.1, AK011334.1, AK019170.1, AK149726.1, AK161290.1, BC058802.1, BC059724.1, NM_001026212.1, NM_019561.2 transcripts.
Set Membership: |
|

Ensa
Genomic Map