Assay Details
| Gene Symbol: Snrpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046637.1 | NP_001040102.1 | 1 | 144 | 88 | |
| GenBank mRNA | AK049541.1 | - | 1 | 538 | 88 | |
| CX565356.1 | - | 1 | 114 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001046637.1 transcripts.
Set Membership: |
|

Snrpa
Genomic Map