Assay Details
| Gene Symbol: Rbm8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102407.1 | NP_001095877.1 | 6 | 2248 | 73 | |
| NM_025875.2 | NP_080151.2 | 6 | 2245 | 73 | ||
| GenBank mRNA | AK134935.1 | - | 1 | 536 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbm8a
Genomic Map